Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159396.1 | NP_001152868.1 | 10 | 1114 | ||
NM_008390.2 | NP_032416.1 | 10 | 1159 | |||
GenBank mRNA | AK152005.1 | - | 10 | 1123 | ||
AK152104.1 | - | 10 | 1122 | |||
AK155983.1 | - | 10 | 1123 | |||
AK157347.1 | - | 10 | 1123 | |||
AY225160.1 | - | 9 | 922 | |||
BC003821.1 | - | 10 | 1092 | 3600525 | ||
CT010234.1 | - | 9 | 890 | |||
M21065.1 | - | 10 | 1095 |