Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133957.1 | - | 10 | 1125 | ||
BC040362.1 | - | 10 | 1186 | 4506177 | ||
BC049100.1 | - | 10 | 1384 | 5698182 | ||
D50366.1 | - | 10 | 1299 | |||
D50367.1 | - | 10 | 1299 |