Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170851.1 | NP_001164322.1 | 9 | 1118 | ||
NM_008462.5 | NP_032488.4 | 8 | 1019 | |||
GenBank mRNA | AF253057.1 | - | 7 | 853 | ||
AF253058.1 | - | 7 | 853 | |||
AF253059.1 | - | 7 | 853 | |||
AF395446.1 | - | 7 | 853 | |||
AK017140.1 | - | 8 | 1010 | |||
AY860971.1 | - | 7 | 853 | |||
BC064711.1 | - | 9 | 1019 | 30295755 | ||
U10304.1 | - | 8 | 947 |