Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013581.3 | NP_038609.3 | 8 | 2178 | ||
GenBank mRNA | AF109377.2 | - | 8 | 2161 | ||
AK031510.1 | - | 7 | 1596 | |||
AK034636.2 | - | 8 | 2183 | |||
AK152464.1 | - | 8 | 2169 | |||
AK158507.1 | - | 8 | 2540 | |||
AK159497.1 | - | 8 | 2180 | |||
AK162971.1 | - | 8 | 2179 | |||
AK171574.1 | - | 8 | 2172 | |||
AK185855.1 | - | 2 | 106 | |||
AK220387.1 | - | 7 | 2622 | |||
BC049541.1 | - | 2 | 628 | |||
BC063056.1 | - | 8 | 2160 | 5720460 |