Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008510.1 | NP_032536.1 | 3 | 350 | ||
GenBank mRNA | AK079820.1 | - | 3 | 363 | ||
BC062249.1 | - | 3 | 350 | 3974601 | ||
D43769.1 | - | 3 | 350 |