Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010751.3 | NP_034881.2 | 1 | 166 | ||
GenBank mRNA | AK036504.1 | - | 1 | 125 | ||
AK151014.1 | - | 1 | 128 | |||
AK151114.1 | - | 1 | 127 | |||
AK151305.1 | - | 1 | 165 | |||
AK151882.1 | - | 1 | 127 | |||
AK153303.1 | - | 1 | 127 | |||
BC034860.1 | - | 1 | 103 | 1448816 | ||
BY106585.1 | - | 1 | 170 | |||
CT010233.1 | - | 1 | 15 | |||
L38926.1 | - | 1 | 51 | |||
U20614.1 | - | 1 | 120 | |||
X83106.1 | - | 1 | 64 |