Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164433.1 | NP_001157905.1 | 15, 16 | 2235 | ||
NM_138315.2 | NP_612188.1 | 16, 17 | 2454 | |||
GenBank mRNA | AK154665.1 | - | 16, 17 | 2394 | ||
AK154930.1 | - | 16, 17 | 2434 | |||
AK159369.1 | - | 15, 16 | 2182 | |||
AK170382.1 | - | 16, 17 | 2394 | |||
AK170827.1 | - | 16, 17 | 2426 | |||
AK170874.1 | - | 16, 17 | 2436 | |||
AK170881.1 | - | 16, 17 | 2420 | |||
AK171234.1 | - | 16, 17 | 2421 | |||
AK171429.1 | - | 16, 17 | 2433 | |||
AK171479.1 | - | 16, 17 | 2427 | |||
AK172562.1 | - | 16, 17 | 2428 | |||
AK172716.1 | - | 16, 17 | 2380 | |||
AK206362.1 | - | 2, 3 | 211 | |||
BC021477.1 | - | 16, 17 | 2402 | 5343552 | ||
BC034682.1 | - | 16, 17 | 2346 | 3485807 |