Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010779.2 | NP_034909.2 | 3 | 338 | ||
GenBank mRNA | AB051900.1 | - | 3 | 286 | ||
AK144385.1 | - | 3 | 339 | |||
AY007569.1 | - | 3 | 338 | |||
BC026198.1 | - | 3 | 314 | 1314753 | ||
CJ046459.1 | - | 3 | 341 | |||
M55617.1 | - | 338 | ||||
X68804.1 | - | 3 | 285 |