Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010781.3 | NP_034911.3 | 2 | 164 | ||
GenBank mRNA | BC024374.1 | - | 2 | 164 | 4983727 | |
L31853.1 | - | 2 | 158 | |||
M57625.1 | - | 2094 | ||||
M57626.1 | - | 3 | 186 | |||
X78542.1 | - | 158 |