Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010806.1 | NP_034936.1 | 11, 12 | 1706 | ||
GenBank mRNA | AK016557.1 | - | 10, 11 | 1825 | ||
CK637291.1 | - | 2, 3 | 358 |