Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010836.3 | NP_034966.1 | 2 | 329 | ||
GenBank mRNA | AB221640.1 | - | 2 | 240 | ||
AK012208.1 | - | 2 | 329 | |||
AK012556.1 | - | 2 | 398 | |||
BC051983.1 | - | 2 | 298 | |||
BC137577.1 | - | 2 | 328 | 8860596 | ||
BC137578.1 | - | 2 | 328 | 8860597 | ||
U62523.1 | - | 2 | 336 |