Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198914.1 | NP_001185843.1 | 7 | 1055 | ||
NM_010848.3 | NP_034978.3 | 7 | 1055 | |||
GenBank mRNA | AK036518.1 | - | 1 | 317 | ||
AK088079.1 | - | 7 | 976 | |||
AK088598.1 | - | 7 | 977 | |||
AK132250.1 | - | 7 | 920 | |||
AK137965.1 | - | 7 | 921 | |||
BC011513.1 | - | 7 | 963 | 3672769 | ||
X02774.1 | - | 7 | 825 |