Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008688.3 | NP_032714.1 | 5 | 867 | ||
NM_026756.2 | NP_081032.2 | 5 | 867 | |||
GenBank mRNA | AF358455.1 | - | 4 | 266 | ||
AF358456.1 | - | 4 | 266 | |||
AF358457.1 | - | 4 | 266 | |||
AF358458.1 | - | 4 | 266 | |||
AK087539.1 | - | 5 | 852 | |||
AK147496.1 | - | 5 | 852 | |||
AK155258.1 | - | 5 | 869 | |||
U57635.1 | - | 5 | 807 | |||
Y07692.1 | - | 5 | 826 | |||
Y07693.1 | - | 5 | 1172 |