Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033217.3 | NP_150086.2 | 4 | 945 | ||
GenBank mRNA | AK037248.1 | - | 4 | 946 | ||
BC038365.1 | - | 4 | 936 | 5367638 |