Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008700.2 | NP_032726.1 | 1 | 522 | ||
GenBank mRNA | AK142146.1 | - | 1 | 427 | ||
BC139299.1 | - | 1 | 368 | 8862321 | ||
BC139303.1 | - | 1 | 368 | 8862325 | ||
L20300.1 | - | 1 | 339 | |||
X75415.1 | - | 1 | 587 |