Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008721.4 | NP_032747.2 | 5, 6 | 843 | ||
GenBank mRNA | AK146466.1 | - | 5, 6 | 732 | ||
BC003320.1 | - | 5, 6 | 810 | 3155595 | ||
BC023887.1 | - | 4, 5 | 877 | 5151868 | ||
L03814.1 | - | 5, 6 | 647 | |||
X67209.1 | - | 5, 6 | 647 |