Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290680.1 | NP_001277609.1 | 5 | 413 | ||
NM_001290681.1 | NP_001277610.1 | 5 | 409 | |||
NM_008741.4 | NP_032767.1 | 5 | 549 | |||
GenBank mRNA | AK011563.1 | - | 4 | 365 | ||
AK075612.1 | - | 5 | 506 | |||
AK088499.1 | - | 5 | 541 | |||
BC018224.1 | - | 5 | 481 | 4527900 | ||
BU058771.1 | - | 5 | 412 | |||
DN176718.1 | - | 5 | 549 | |||
U17259.1 | - | 5 | 509 |