Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013614.2 | NP_038642.2 | 9 | 1262 | ||
GenBank mRNA | AK077679.1 | - | 9 | 1164 | ||
AK139610.1 | - | 9 | 1164 | |||
AK150283.1 | - | 9 | 1164 | |||
AK150343.1 | - | 9 | 1163 | |||
AK152525.1 | - | 9 | 1162 | |||
AK166093.1 | - | 9 | 1169 | |||
AK168219.1 | - | 9 | 1163 | |||
AK169214.1 | - | 9 | 1162 | |||
AK172568.1 | - | 9 | 1164 | |||
BC033264.1 | - | 9 | 1153 | 2645291 | ||
BC059826.1 | - | 9 | 1143 | 6808137 | ||
BC083122.1 | - | 9 | 1150 | 6329274 | ||
EU684749.1 | - | 7 | 852 | |||
M12330.1 | - | 4 | 321 | |||
M20617.2 | - | 7 | 868 | |||
S64539.1 | - | 9 | 1162 |