Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252565.1 | NP_001239494.1 | 3 | 549 | ||
NM_011012.5 | NP_035142.1 | 3 | 621 | |||
GenBank mRNA | AF043276.1 | - | 4 | 706 | ||
AF043277.1 | - | 3 | 492 | |||
AF043278.1 | - | 3 | 533 | |||
AF062381.1 | - | 2 | 395 | |||
AF075605.1 | - | 2 | 394 | |||
AF126469.1 | - | 2 | 379 | |||
AK031926.1 | - | 4 | 733 | |||
AK038620.1 | - | 3 | 551 | |||
AK043275.1 | - | 4 | 605 | |||
AK044178.1 | - | 4 | 575 | |||
AK079529.1 | - | 3 | 594 | |||
BC050885.1 | - | 3 | 543 | 5703258 | ||
BC051982.1 | - | 3 | 481 | 5695055 | ||
U04952.1 | - | 3 | 542 | |||
U09421.1 | - | 3 | 672 | |||
U14165.1 | - | 2 | 382 | |||
X91813.1 | - | 3 | 552 |
Set Membership: |
G-protein coupled receptor |