Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013623.2 | NP_038651.2 | 4, 5 | 476 | ||
GenBank mRNA | BC141235.1 | - | 4, 5 | 480 | 9055847 | |
BC145559.1 | - | 4, 5 | 480 | 9054123 | ||
BC145560.1 | - | 4, 5 | 510 | |||
BC171932.1 | - | 3, 4 | 337 |