Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008786.2 | NP_032812.2 | 6, 7 | 962 | ||
GenBank mRNA | AK006162.1 | - | 6, 7 | 640 | ||
AK075632.1 | - | 5, 6 | 449 | |||
AK208253.1 | - | 1, 2 | 95 | |||
AK214928.1 | - | 3, 4 | 229 | |||
BC040750.1 | - | 6, 7 | 555 | |||
BC049613.1 | - | 6, 7 | 666 | |||
BC058966.1 | - | 6, 7 | 651 | 5717963 | ||
M60320.1 | - | 6, 7 | 626 |