Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008801.2 | NP_032827.1 | 3 | 329 | ||
GenBank mRNA | AF039216.1 | - | 3 | 288 | ||
AK141209.1 | - | 3 | 335 | |||
AK165701.1 | - | 3 | 554 | |||
AK170067.1 | - | 3 | 341 | |||
BC005636.1 | - | 3 | 329 | 3964336 |