Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pemt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290011.1 | NP_001276940.1 | 5 | 550 | ||
NM_001290012.1 | NP_001276941.1 | 5 | 546 | |||
NM_001290013.1 | NP_001276942.1 | 5 | 484 | |||
NM_008819.3 | NP_032845.2 | 5 | 584 | |||
GenBank mRNA | AK133744.1 | - | 5 | 546 | ||
AY334571.1 | - | 5 | 443 | |||
BC026796.1 | - | 5 | 418 | 4196324 | ||
BG084478.2 | - | 5 | 448 | |||
CJ048474.1 | - | 5 | 546 |