Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278581.1 | NP_001265510.1 | 3 | 386 | ||
NM_001278582.1 | NP_001265511.1 | 3 | 321 | |||
NM_001278583.1 | NP_001265512.1 | 3 | 291 | |||
NM_001278584.1 | NP_001265513.1 | 2 | 180 | |||
NM_008895.4 | NP_032921.1 | 2 | 210 | |||
GenBank mRNA | AK017492.1 | - | 2 | 206 | ||
AK017581.1 | - | 2 | 175 | |||
AK030714.1 | - | 3 | 285 | |||
AK077426.1 | - | 2 | 174 | |||
AK133592.1 | - | 2 | 205 | |||
AK133646.1 | - | 2 | 205 | |||
AK133740.1 | - | 2 | 204 | |||
AK133776.1 | - | 2 | 206 | |||
AK133800.1 | - | 2 | 206 | |||
BC061215.1 | - | 2 | 204 | 30253829 |