Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Procr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011171.2 | NP_035301.2 | 3 | 669 | ||
GenBank mRNA | AK167302.1 | - | 3 | 609 | ||
BC028755.1 | - | 3 | 542 | 1054063 | ||
CT010175.1 | - | 3 | 488 | |||
L39017.1 | - | 3 | 543 | |||
X63748.2 | - | 3 | 575 |