Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008944.2 | NP_032970.2 | 4, 5 | 408 | ||
GenBank mRNA | AF099733.1 | - | 4, 5 | 375 | ||
AK012119.1 | - | 4, 5 | 390 | |||
AK012990.1 | - | 4, 5 | 397 | |||
AK035578.1 | - | 397 | ||||
AK136115.1 | - | 4, 5 | 410 | |||
AK151883.1 | - | 4, 5 | 399 | |||
AK152862.1 | - | 4, 5 | 399 | |||
AK154011.1 | - | 4, 5 | 392 | |||
AK167752.1 | - | 4, 5 | 399 | |||
BC026768.1 | - | 4, 5 | 381 | 3156907 | ||
CJ052982.1 | - | 4, 5 | 410 | |||
X70303.1 | - | 4, 5 | 452 |