Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011188.3 | NP_035318.1 | 2 | 386 | ||
GenBank mRNA | AK076874.1 | - | 2 | 388 | ||
AK146919.1 | - | 1 | 27 | |||
AK153413.1 | - | 1 | 30 | |||
AK168548.1 | - | 1 | 27 | |||
BC005462.1 | - | 1 | 36 | 3590739 |