Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016676.5 | NP_057885.1 | 3, 4 | 876 | ||
GenBank mRNA | AB232604.1 | - | 3, 4 | 325 | ||
AF035646.1 | - | 3, 4 | 566 | |||
AK028320.1 | - | 3, 4 | 847 | |||
AK087964.1 | - | 3, 4 | 838 | |||
AK134988.1 | - | 3, 4 | 796 | |||
AK146357.1 | - | 3, 4 | 877 | |||
AK150319.1 | - | 3, 4 | 836 | |||
AK150384.1 | - | 3, 4 | 530 | |||
AK150791.1 | - | 3, 4 | 826 | |||
AK151473.1 | - | 3, 4 | 530 | |||
AK153315.1 | - | 3, 4 | 836 | |||
AK153325.1 | - | 3, 4 | 826 | |||
AK159383.1 | - | 3, 4 | 841 | |||
AK159845.1 | - | 3, 4 | 841 | |||
AK159988.1 | - | 3, 4 | 835 | |||
AK160960.1 | - | 3, 4 | 831 | |||
AK168320.1 | - | 3, 4 | 836 | |||
AK190935.1 | - | 2, 3 | 178 | |||
AK195457.1 | - | 3, 4 | 312 | |||
AK202554.1 | - | 1, 2 | 168 | |||
AK212462.1 | - | 1, 2 | 110 | |||
BC052735.1 | - | 826 | ||||
BC056374.1 | - | 3, 4 | 827 | 6848840 | ||
CT010394.1 | - | 3, 4 | 325 |