Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159725.2 | NP_001153197.2 | 4 | 548 | ||
NM_008998.4 | NP_033024.1 | 4 | 779 | |||
GenBank mRNA | AB232611.1 | - | 3 | 385 | ||
AK009707.1 | - | 4 | 549 | |||
AK075591.1 | - | 4 | 780 | |||
BC013170.1 | - | 4 | 628 | 2648145 | ||
BC051071.1 | - | 4 | 755 | 6307319 | ||
X70804.1 | - | 3 | 385 |