Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfxank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025589.1 | NP_001020760.1 | 4 | 594 | ||
NM_011266.2 | NP_035396.1 | 4 | 594 | |||
GenBank mRNA | AF094761.1 | - | 4 | 510 | ||
AF123704.1 | - | 4 | 495 | |||
AK052298.1 | - | 4 | 551 | |||
AK146148.1 | - | 4 | 523 | |||
AK162595.1 | - | 1 | 38 | |||
BC010971.1 | - | 4 | 528 | 4190691 | ||
CN526109.1 | - | 4 | 594 | |||
L43164.1 | - | 4 | 495 |