Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sord | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146126.4 | NP_666238.1 | 3 | 265 | ||
GenBank mRNA | AK004692.1 | - | 3 | 289 | ||
AK015059.1 | - | 3 | 266 | |||
AK152169.1 | - | 3 | 314 | |||
AK166988.1 | - | 3 | 265 | |||
AK166996.1 | - | 3 | 294 | |||
AK178689.1 | - | 1 | 77 | |||
AK209251.1 | - | 2 | 151 | |||
BC024124.1 | - | 3 | 280 | 5099395 | ||
BC030875.1 | - | 3 | 254 | 4237164 | ||
BC039957.1 | - | 3 | 265 | |||
BC092291.1 | - | 3 | 268 | 5097505 | ||
U27014.1 | - | 265 |