Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009149.2 | NP_033175.1 | 3 | 532 | ||
GenBank mRNA | AK154581.1 | - | 3 | 547 | ||
BC021306.1 | - | 3 | 543 | 4239405 | ||
X84037.1 | - | 3 | 532 |