Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011381.4 | NP_035511.2 | 1 | 867 | ||
GenBank mRNA | BC094426.1 | - | 670 | 6855602 | ||
BC098096.1 | - | 1 | 487 | 6402835 | ||
BF467709.1 | - | 1 | 505 | |||
D83144.1 | - | 1 | 1128 | |||
D83145.1 | - | 1 | 453 | |||
X90871.1 | - | 642 |