Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133854.3 | NP_598615.1 | 3 | 320 | ||
GenBank mRNA | AF086838.1 | - | 3 | 281 | ||
AK007458.1 | - | 3 | 301 | |||
AK035147.1 | - | 3 | 317 | |||
AK044085.1 | - | 2 | 254 | |||
AK048007.1 | - | 3 | 309 | |||
AK050151.1 | - | 3 | 321 | |||
AK077840.1 | - | 3 | 311 | |||
AK080718.1 | - | 3 | 311 | |||
AK086994.1 | - | 2 | 252 | |||
AK135006.1 | - | 3 | 312 | |||
AK152057.1 | - | 3 | 289 | |||
BC006744.1 | - | 1 | 247 | 3708620 | ||
BC048691.1 | - | 3 | 292 | 6511395 |