Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166480.1 | NP_001159952.1 | 6 | 666 | ||
NM_009279.3 | NP_033305.1 | 6 | 548 | |||
GenBank mRNA | AK003044.1 | - | 6 | 528 | ||
AK007921.1 | - | 6 | 504 | |||
AK013723.1 | - | 6 | 510 | |||
AK142600.1 | - | 1 | 3571 | |||
AK160089.1 | - | 6 | 514 | |||
AK207479.1 | - | 2 | 83 | |||
AK214704.1 | - | 2 | 83 | |||
BC008172.1 | - | 6 | 469 | 3481948 | ||
BC012971.1 | - | 3 | 2443 | |||
BC019419.1 | - | 5 | 1999 | |||
BQ747160.1 | - | 6 | 451 | |||
X90582.1 | - | 6 | 629 |