Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139152.1 | NP_631891.1 | 3 | 479 | ||
GenBank mRNA | AY057054.1 | - | 3 | 479 | ||
BC107370.1 | - | 2 | 356 | |||
BC127152.1 | - | 2 | 356 | |||
BC141295.1 | - | 2 | 445 | 9055907 | ||
BC145621.1 | - | 2 | 445 | 9054189 |