Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164493.1 | NP_001157965.1 | 8 | 2163 | ||
GenBank mRNA | AK122572.1 | - | 8 | 1816 | ||
BC065168.1 | - | 8 | 2163 | |||
BC138283.1 | - | 7 | 1519 | 8861302 | ||
BC145230.1 | - | 7 | 1519 | |||
BC145748.1 | - | 7 | 1519 | 40131051 |