Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105196.1 | NP_001098666.1 | 9 | 1513 | ||
NM_001105197.1 | NP_001098667.1 | 10 | 1304 | |||
NM_001271489.1 | NP_001258418.1 | 10 | 1599 | |||
NM_001271490.1 | NP_001258419.1 | 10 | 1771 | |||
NM_001271491.1 | NP_001258420.1 | 9 | 1233 | |||
NM_172472.3 | NP_766060.2 | 10 | 1618 | |||
GenBank mRNA | AK089762.1 | - | 8 | 1275 | ||
AK139756.1 | - | 9 | 1510 | |||
AK144657.1 | - | 10 | 1583 | |||
AK144751.1 | - | 10 | 1587 | |||
AK145029.1 | - | 10 | 1739 | |||
AK145832.1 | - | 10 | 1583 | |||
AK147405.1 | - | 7 | 4677 | |||
AK150372.1 | - | 10 | 1370 | |||
AK151447.1 | - | 10 | 1289 | |||
AK152221.1 | - | 10 | 1289 | |||
AK153051.1 | - | 10 | 1370 | |||
AK154690.1 | - | 8 | 3073 | |||
AK155883.1 | - | 9 | 1235 | |||
AK159162.1 | - | 10 | 1498 | |||
AK196916.1 | - | 1 | 62 | |||
BC056358.1 | - | 10 | 1606 | 6813486 | ||
BC063047.1 | - | 10 | 1606 | 6813486 | ||
S76673.1 | - | 8 | 956 |