Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009302.3 | NP_033328.3 | 3, 4 | 525 | ||
GenBank mRNA | AF053974.1 | - | 3, 4 | 417 | ||
AK019882.1 | - | 3, 4 | 469 | |||
AK041505.1 | - | 3, 4 | 473 | |||
AK045295.1 | - | 3, 4 | 497 | |||
AK146773.1 | - | 3, 4 | 517 | |||
AK172990.1 | - | 3, 4 | 462 | |||
BC065136.1 | - | 3, 4 | 467 | 30546814 |