Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081329.1 | NP_001074798.1 | 7 | 2705 | ||
GenBank mRNA | AK034954.1 | - | 7 | 2707 | ||
BC028272.1 | - | 1 | 431 | |||
BC054371.1 | - | 1 | 420 |