Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081278.2 | NP_001074747.2 | 11 | 2825 | ||
GenBank mRNA | AK031120.1 | - | 11 | 2669 | ||
AK039337.1 | - | 13 | 3015 | |||
AK044719.1 | - | 11 | 2467 | |||
AK220347.1 | - | 10 | 1232 | |||
BC042747.1 | - | 7 | 827 | |||
BC064433.1 | - | 11 | 2449 | |||
BC080762.1 | - | 11 | 2443 | |||
BC117869.1 | - | 11 | 2298 | 40095526 | ||
BC117870.1 | - | 12 | 2325 | 40095531 |