Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Churc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206534.1 | NP_996257.1 | 4 | 313 | ||
GenBank mRNA | BC019472.1 | - | 3 | 240 | ||
BC076499.1 | - | 4 | 313 | 6815558 |