Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172503.3 | NP_766091.2 | 14 | 3110 | ||
GenBank mRNA | AK052168.1 | - | 14 | 3124 | ||
AK053663.1 | - | 14 | 3124 | |||
AK131119.1 | - | 9 | 1968 | |||
AK155465.1 | - | 14 | 3124 | |||
BC016518.1 | - | 4 | 929 | |||
BC058080.1 | - | 14 | 3110 | 6822376 |