Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033247.1 | NP_001028419.1 | 9 | 1129 | ||
GenBank mRNA | AK078103.1 | - | 4 | 821 |