Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130457.1 | NP_001123929.1 | 3 | 617 | ||
NM_134011.2 | NP_598772.1 | 3 | 617 | |||
GenBank mRNA | AK028966.1 | - | 3 | 595 | ||
AK144813.1 | - | 3 | 615 | |||
AK166760.1 | - | 4 | 525 | |||
AK168088.1 | - | 3 | 615 | |||
BC016483.1 | - | 3 | 573 | 3155271 | ||
BC031155.1 | - | 3 | 588 | 4976690 | ||
BC031910.1 | - | 3 | 587 | 5362943 |