Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289603.1 | NP_001276532.1 | 9 | 979 | ||
NM_033476.3 | NP_258437.1 | 9 | 979 | |||
GenBank mRNA | AK141248.1 | - | 3 | 319 | ||
AK146503.1 | - | 9 | 980 | |||
AK147898.1 | - | 8 | 1188 | |||
AK158682.1 | - | 9 | 899 | |||
BC022131.1 | - | 9 | 952 | 5098336 | ||
M84809.1 | - | 9 | 918 |