Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162868.1 | NP_001156340.1 | 12 | 2755 | ||
NM_001162869.1 | NP_001156341.1 | 13 | 2890 | |||
NM_153140.2 | NP_694780.1 | 10 | 1146 | |||
GenBank mRNA | AB093257.1 | - | 13 | 2331 | ||
AK083658.1 | - | 12 | 2440 | |||
AK085516.1 | - | 10 | 1147 | |||
AK128985.1 | - | 10 | 1146 | |||
BC023364.1 | - | 10 | 1137 | |||
BC037132.1 | - | 10 | 1186 | |||
CB840427.1 | - | 1 | 41 |