Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172550.4 | NP_766138.3 | 3 | 652 | ||
GenBank mRNA | AK037727.1 | - | 3 | 379 | ||
AK136543.1 | - | 3 | 403 | |||
BC057308.1 | - | 3 | 652 | 5703033 |