Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195268.1 | NP_001182197.1 | 9 | 1446 | ||
GenBank mRNA | AK035824.1 | - | 3 | 597 | ||
AK137009.1 | - | 11 | 1783 | |||
BC024795.1 | - | 1 | 673 | |||
BC026893.1 | - | 1 | 887 | |||
BC039779.1 | - | 1 | 610 | |||
BC052075.1 | - | 10 | 1500 | |||
BC078442.1 | - | 5 | 1001 | |||
BC100552.1 | - | 4 | 829 | 5687195 |