Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285785.1 | NP_001272714.1 | 13, 14 | 1684 | ||
NM_146011.2 | NP_666123.2 | 12, 13 | 1123 | |||
GenBank mRNA | AK047280.1 | - | 12, 13 | 1068 | ||
AK049245.1 | - | 9, 10 | 904 | |||
AK083474.1 | - | 9, 10 | 1033 | |||
AK089051.1 | - | 12, 13 | 1075 | |||
AK167738.1 | - | 13, 14 | 1261 | |||
BC021754.1 | - | 2, 3 | 98 | |||
BC024535.1 | - | 12, 13 | 1118 | 4987970 | ||
BC027374.1 | - | 12, 13 | 1094 | 4947306 | ||
DQ498128.1 | - | 13, 14 | 1532 |